Learn More
Lenz Laborglasinstrumente™ Splash Heads Acc. to Stutzer, Straight
For placement onto distillation flasks for separation of drops. Lenz Laborglasinstrumente™ Splash Heads Acc. to Stutzer, Straight, are manufactured from durable Duran™ glass tubing.
£64.00 - £87.00
Specifications
For Use With (Application) | For separation of drops (Direct placement onto distillation flasks) |
---|---|
Type | Straight Splash Heads |
Material | Duran Glass |
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
10588421
|
Lenz Laborglasinstrumente™
05035011 |
NS 14/23 NS 14/23 |
£64.00
Each |
Estimated Shipment: 14-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11953461
|
Lenz Laborglasinstrumente™
05035045 |
NS 29/32, NS 14/23 |
£87.00
Each |
Estimated Shipment: 21-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10231071
|
Lenz Laborglasinstrumente™
05035044 |
NS 29/32, NS 29/32 |
£86.66
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Straight with NS cone and socket
- Temperature and chemical resistance
- Minimal thermal expansion
Specifications
For separation of drops (Direct placement onto distillation flasks) | |
Straight Splash Heads | |
Duran Glass |