missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ Straight Suction Tube for Buerkle™ ProfiSampler
These disposable tubes can be immersed absolutely vertically into the container.
£146.00 - £198.00
Specifications
Product Type | Suction Tube |
---|---|
Style | Straight |
Weight (Metric) | 0.8 kg |
Diameter (Metric) Outer | 6 x 8 mm |
Product Code | Brand | Color | For Use With (Application) | Length (Metric) | Material | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | For Use With (Application) | Length (Metric) | Material | Price | Quantity & Availability | ||
11862303
|
Buerkle™
5305-1024 |
Black | For Flammable Liquids in VbF Hazard Category AI (Electrically Conducting) | 220 cm | Polyamide |
£198.00
Pack of 10 |
In Stock
Log in to see stock. |
||
11892283
|
Buerkle™
5305-1022 |
Transparent | For General Sampling | 200 cm | Polyethylene |
£146.00
Pack of 10 |
Estimated Shipment: 29-07-2024
Log in to see stock. |
||
Description
- Manufactured from either polyethylene or polyamide plastic
- Polyethylene tubes used for general sampling, polyamide tubes used for flammable liquids in VbF Hazard Category AI (electrically conducting)
- Diameter of 6 x 8mm
- Not autoclavable
Specifications
Suction Tube | |
Straight | |
0.8 kg | |
6 x 8 mm |