Learn More
Lenz Laborglasinstrumente™ Straight Tubing Connectors
Manufactured from DURAN™ Lenz Laborglasinstrumente™ Straight Tubing Connectors are ideal for connecting tubes of various diameters.
Brand: Lenz Laborglasinstrumente™ 01600010
Additional Details : Weight : 0.10000kg
Description
- Tubing connectors
- Straight
- With two glass olives
- Made of DURAN™ tubing
Specifications
10 mm | |
115 mm | |
10 |