missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences Strep-Tactin™ Superflow™, 50% Suspension

Used for purification of recombinant Strep-tag proteins under gravity flow and low pressure or FPLC

£100.20 - £1295.00

Specifications

Stability 6 months after shipping.
Storage Requirements 2°C to 8°C
Binding Capacity Biotin: >300 nmol/ mL resin
Product Type 50% suspension
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12846446
View Documents Promotion Details
IBA Lifesciences
2-1206-002
4 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12866446
View Documents Promotion Details
IBA Lifesciences
2-1206-025
50 mL
£1295.00
50mL
View alternative products
Save Up to 
»
Estimated Shipment: 16-10-2024
to see stock.
Add to basket
 
Description

Description

  • High capacity resin provides 2 to 3 times higher binding capacity for strep-tag proteins than standard Strep-Tactin Superflow suspension
  • Higher yields of highly pure Strep-tag protein
  • Bulk resin is recommended for packing of columns and cartridges since the resin is optimized for column affinity chromatography rather than for batch purification
  • Binding capacity: 1mL sedimented resin (corresponding to 2mL of 50 % suspension) is useful for one-step purification of 50 to 100nmol recombinant Strep-tag protein (up to 3mg in case of  30 kDa protein)
Specifications

Specifications

6 months after shipping.
2°C to 8°C
Biotin: >300 nmol/ mL resin
50% suspension
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.