missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Strep-Tactin™ XT Sepharose™ Chromatography Resin
Chromatography resin for purification of recombinant proteins tagged with a Strep-tag™ II and Twin-Strep-tag™.
£891.28 - £3980.00
Specifications
For Use With (Application) | Purification of recombinant proteins tagged with Strep-tag II and Twin-Strep-tag |
---|---|
Stability | Stable in all commonly used aqueous buffers |
Storage Requirements | 2 to 8°C in 20% ethanol |
Binding Capacity | Approx. 10 mg protein with Strep-tag II or Twin-Strep-tag/mL of resin |
Particle Size | 34 μm |
Product Code | Brand | Packaging Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging Quantity | Price | Quantity & Availability | |||||
16486149
|
Cytiva
29401324 |
10 mL |
£891.28
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16496149
|
Cytiva
29401326 |
50 mL |
£3980.00
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Highly pure Strep-tag II and Twin-Strep-tag recombinant proteins
- Fast purifications with simple elution
- Physiological conditions and mild elution preserve target protein activity
- Fast and easy regeneration
- Compatible with a wide range of detergents, denaturants, and other additives
Specifications
Purification of recombinant proteins tagged with Strep-tag II and Twin-Strep-tag | |
2 to 8°C in 20% ethanol | |
34 μm | |
6 to 10 pH | |
Strep-Tactin XT |
Stable in all commonly used aqueous buffers | |
Approx. 10 mg protein with Strep-tag II or Twin-Strep-tag/mL of resin | |
Rigid, cross-linked agarose, spherical | |
Approx. 5 mg/mL resin |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- StrepTactin XT Sepharose, 10 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva Strep-Tactin™ XT Sepharose™ Chromatography Resin