missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences 10x Buffer W - Wash Buffer

Strep-Tactin™ and Strep-Tactin™XT wash buffer

Brand:  IBA Lifesciences 2-1003-100

Code : LE

Additional Details : Weight : 0.13140kg


Product Code. 12813206

  • £70.35 / 100mL
Estimated Shipment: 06-08-2024
to see stock.

Description

Description

  • 10x Buffer W is a 10x concentrated wash buffer for Strep-Tactin™ and Strep-Tactin™XT resins.
  • It removes unbound molecules from the affinity chromatography matrix during Strep-tag™II and Twin-Strep-tag™ fusion protein purification.
Specifications

Specifications

Wash Buffer
100 mL
For Removal of Unbound Molecules From Immobilized Strep-Tactin During Strep-Tag Protein Purification
10 X
Washing Buffer
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Buffer W (10x), Strep-Tactin/Strep-Tactin XT Wash Buffer

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Tris, EDTA (Ethylenediaminetetraacetic acid)

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.