missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences StrepMAB-Classic

Monoclonal antibody to detect Strep-tag™II and Twin-Strep-tag™ fusion proteins
Brand: IBA Lifesciences 2-1507-001
This item is not returnable.
View return policy
Description
StrepMAB-Classic is a highly selective, murine monoclonal antibody for detection of Strep-tag™II and Twin-Strep-tag™ fusion proteins in applications like western blots, ELISA, immunohistochemistry, and immunocytochemistry.It is available unconjugated or coupled to horseradish peroxidase (HRP). StrepMAB-Classic HRP is suitable for chromogenic and chemiluminescent reactions. Please note: For capturing of Strep-tag™II and Twin-Strep-tag™ fusion proteins on solid phases, the use of StrepMAB-Immo or Strep-Tactin™XT is recommended due to the binding affinity in the pM range.Specifications
Monoclonal Antibody | |
2°C to 8°C | |
100 μg |
RUO | |
Suspension in PBS |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Protocol Western-blot 2-1507-001
File size: 629.5 kB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IBA Lifesciences StrepMAB-Classic > Product Type: not conjugated
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction