missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Horizon™ APC-R700 Streptavidin
APC-R700 Streptavidin in a non-glycosylated protein that is prepared chromatographically from the bacterium Streptomyces avidinii. APC-R700 exhibits significantly improved brightness.
Brand: BD 565144
Additional Details : Weight : 0.01000kg
Description
- For flow cytometry, fluorescence microscopy and imaging.
- Exhibits significantly improved brightness.
Specifications
Streptavidin | |
APC-R700 | |
For research use only (RUO) | |
Store undiluted at 4°C. Protect from prolonged exposure to light. Do not freeze. |
0.1 mg/mL | |
0.1 mg | |
Aqueous buffered solution containing protein stabilizer and less than or equal to 0.09% sodium azide | |
Flow Cytometry |