missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Streptavidin, BV711
100μg in 0.1mg/mL concentration
Brand: BD 563262
Additional Details : Weight : 0.02000kg
Description
Host Species:
Species Reactivity:Ms
Flow Cytometry
Specifications
0.1 mg/mL | |
100 μg | |
4°C | |
Flow Cytometry |