missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Streptavidin eFluor™ 615 Conjugate
Commonly used in indirect staining protocols to detect biotinylated primary antibodies in immunohistochemistry and immunocytochemistry applications. Streptavidin binds to biotin with high affinity.
£243.00 - £370.00
Specifications
Concentration | 0.2 mg/mL |
---|---|
Conjugate | eFluor 615 |
Form | Liquid |
Product Line | eBioscience™ |
Product Type | Streptavidin Conjugate (fluorescent) |
Alert:
Contains Sodium azide
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15351100
|
Invitrogen™
42-4317-80 |
25 μg |
£243.00
25µg |
Estimated Shipment: 06-12-2024
Log in to see stock. |
|||||
15361100
|
Invitrogen™
42-4317-82 |
100 μg |
£370.00
100µg |
Estimated Shipment: 06-12-2024
Log in to see stock. |
|||||
Description
Specifications
0.2 mg/mL | |
Liquid | |
Streptavidin Conjugate (fluorescent) |
eFluor 615 | |
eBioscience™ | |
Biotin |
For Research Use Only