Learn More
Molecular Probes™ Streptavidin, horseradish peroxidase conjugate
The streptavidin, horseradish peroxidase (HRP) conjugate can be used to detect biotin in a signal amplification scheme in conjunction with chromogenic substrates or fluorogenic HRP substrates.
Brand: Molecular Probes™ S911
172.92 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Description
The streptavidin, horseradish peroxidase (HRP) conjugate can be used to detect biotin in a signal amplification scheme in conjunction with chromogenic substrates or fluorogenic HRP substrates, such as the tyramide signal amplification (TSA) technology.
We recommend reconstituting in 1 mL of 1X phosphate-buffered saline (PBS), pH 7.2, for a 1 mg/mL stock solution.
Specifications
HRP | |
Store in freezer (-5°C to -30°C). | |
Streptavidin Conjugate (fluorescent) |
1 mg | |
Wet Ice | |
HRP |
Safety and Handling
- STREPTAVIDIN, HORSERADISH
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H332-Harmful if inhaled.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P314-Get medical advice/attention if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : thimerosal
For Research Use Only. Not for use in diagnostic procedures.