missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Streptavidin PerCP-Cyanine5.5 Conjugate
Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.
£124.00 - £209.00
Specifications
Concentration | 0.2 mg/mL |
---|---|
Conjugate | PerCP-Cy5.5 |
Form | Liquid |
Product Line | eBioscience™ |
Product Type | Streptavidin Conjugate (Fluorescent) |
Alert:
Contains Sodium azide
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15311180
|
Invitrogen™
45-4317-80 |
25 μg |
£124.00
25µg |
Estimated Shipment: 07-10-2024
Log in to see stock. |
|||||
15321180
|
Invitrogen™
45-4317-82 |
100 μg |
£209.00
100µg |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||||
Description
Specifications
0.2 mg/mL | |
Liquid | |
Streptavidin Conjugate (Fluorescent) | |
PerCP-Cy5.5 |
PerCP-Cy5.5 | |
eBioscience™ | |
Biotin |
For Research Use Only