missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Streptavidin Protein, AP
Protein
Brand: Invitrogen™ 21323
Code : PN
Additional Details : Weight : 0.80000kg
Description
Streptavidin Protein, AP conjugate, 21323, from Invitrogen., 21323, from Invitrogen. Species Reactivity: ; Applications: ELISA, Immunofluorescence, Western Blot.
Streptavidin is biotin-binding protein that was originally isolated from Streptomyces avidinii. In contrast to avidin, streptavidin has no carbohydrate and has a mildly acidic pI of 5. Thermo Scientific Streptavidin products use a recombinant form of streptavidin having a mass of 53,000 daltons and a near-neutral pI. Streptavidin is a tetrameric protein, with each subunit binding one molecule of biotin with affinity similar to that of avidin. Guanidinium chloride will dissociate avidin and streptavidin into subunits, but streptavidin is more resistant to dissociation.Specifications
2-4mg/mL | |
Tag | |
8 | |
RUO | |
Liquid | |
purified | |
Streptavidin |
AP | |
TBS with 0.1mM MgCl2, 0.1mM ZnCl2, 50% glycerol and no preservative; pH 8 | |
3mg | |
-20°C | |
ELISA, Western Blot | |
N/A |