Learn More
Molecular Probes™ Streptavidin, Unconjugated
Streptavidin, Unconjugated
Brand: Molecular Probes™ SNN1001
Code : 50
Additional Details : Weight : 0.05250kg
Description
Purified from Streptomyces avidinii and provided in lyophilized form. The streptavidin/avidin-biotin staining and detection strategy is used when increased assay sensitivity is desired for a wide variety of systems.Format/Formulation: Unconjugated
Specifications
Unconjugated | |
Wet Ice | |
Streptavidin Conjugate (fluorescent) |
Store in refrigerator (2–8°C). | |
Solid |
For Laboratory Use