missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Streptex™ Latex Agglutination Test

Product Code. p-6100325
Click to view available options
Quantity:
200 Tests/Kit
50 Tests/Kit
Unit Size:
200 tests
50 tests
This item is not returnable. View return policy

Product Code. 11434099

Brand: Thermo Scientific™ R30950501

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate and identify a variety of streptococci with a simple enzyme extraction method that detects Lancefield groups A, B, C, D, F and G.

Rapidly identify and differentiate Lancefield group of streptococci using the Thermo Scientific™ Streptex™ Latex Agglutination Test. The test utilizes latex particles sensitized with group-specific antibodies which agglutinate in presence of homologous antigens.

Product Features
Use this rapid latex agglutination test, providing a complete solution for the isolation and differentiation of Lancefield groups A, B, C, D, F & G.

View distinct, clear results with white latex agglutination on a black background. Individual latex grouping reagents are available as required.

With Streptex, group specific antigens are extracted from streptococci in a simple incubation step. Antigens are then identified using polystyrene latex particles which have been coated with group-specific antibodies. These latex particles agglutinate strongly in the presence of homologous antigen and remain in smooth suspension in the absence of homologous antigen.

For a more rapid acid extraction method, Streptex Acid Extraction Kit (Part No. R30951301) is available for use with Streptex. Acid extraction will rapidly extract the Group antigens without the need for incubation. However, Group D antigen is not extracted efficiently with acid extraction; therefore, an alternative method must be used to confirm suspected Group D streptococci.

Proven Performance

  • Rely on tried-and-true tests with more than 50 years of combined proven performance
  • Maintain confidence with reliable results that have greater than 99% sensitivity (according to data available in the package inserts)

Workflow Efficiency

  • Clear-cut, robust reactions provide easy-to-read results
  • Improve productivity with compact kit configurations, color-coded reagents and convenient workstations
  • Flexible use from selective and non-selective media
  • Control latex is not required

Economical Solution

  • Maximize spending efficiency with the option of purchasing separate kit components
  • Save time, money, and resources by turning to one company, one source, for a complete set of Staph identification solutions

Streptococci are facultatively anaerobic, catalase-negative, gram-positive cocci that occur as pairs or chains. They cause a wide variety of clinical diseases including pharyngitis, scarlet fever, otitis media, impetigo, meningitis, neonatal sepis, pneumonia, endocarditis, osteomyelitis, and necrotizing fascitis. One of the most useful phenotypic characteristics of streptococci is the hemolytic reaction, generally classified as alpha, beta, or gamma, according to the appearance of zones around colonies growing on Blood Agar. Beta-hemolytic streptococci are further characterized by Lancefield groups based on the antigenic differences in group-specific polysaccharides located in the bacterial cell wall.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Control Sets Positive
Description Streptex Latex Agglutination Test
CE Marker Yes
Detectable Analytes Streptococci (Group A, B, C, F, G)
Type Streptococcus Test Kit
For Use With (Application) Enzyme Immunoassay Detection
Includes Test latex, extraction enzyme, polyvalent positive control, disposable reaction cards, disposable mixing sticks
Material Polyvalent
Organism Group Lancefield A, B, C, D, F
Quantity 50 Tests/Kit
Target Organism Class Streptococcus
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.