missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Streptomycin sulfate Product Code.: 15424871

Thermo Scientific Chemicals Streptomycin sulfate

Product Code. 15424871
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 15424871

Brand: Thermo Scientific Chemicals 455340250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Streptomycin sulfate, CAS # 3810-74-0, is the sulfate salt form of streptomycin, and an aminoglycoside with antibacterial activity. It also shows a protein synthesis inhibitory activity.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Streptomycin sulfate is an aminoglycoside sulfate salt derived from Streptomyces griseus that shows antibacterial activity.

• It inhibits initiation and causes misreading of rRNA in protein synthesis. This compound binds to the S12 protein of the bacterial 30S ribosomal subunit, thereby inhibiting peptide elongation and protein synthesis.

Application

• Streptomycin sulfate can be used to study the mechanisms of streptomycin resistance

• This compound can be used in laboratory applications combined with penicillin to inhibit bacterial contamination in in vitro cell culture

• Streptomycin sulfate shows antibiotic activity against Mycobacterium tuberculosis

• This compound can be used as a protein synthesis inhibitor

• It acts against gram-negative and gram-positive bacteria • Streptomycin sulfate also shows a fungicide activity

• It can be used to remove nucleic acids from a biological extract

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3810-74-0
Molecular Formula C42H84N14O36S3
Molecular Weight (g/mol) 1457.376
InChI Key QTENRWWVYAAPBI-FFCQDDOVSA-N
PubChem CID 124080941
IUPAC Name 2-[(1R,2S,3S,4S,5R,6S)-3-(diaminomethylideneamino)-4-[(2R,3S,4S,5S)-3-[(2R,3S,4R,5R,6S)-4,5-dihydroxy-6-(hydroxymethyl)-3-(methylamino)oxan-2-yl]oxy-4-formyl-4-hydroxy-5-methyloxolan-2-yl]oxy-2,5,6-trihydroxycyclohexyl]guanidine;2-[(1R,2S,3S,4R,5R,6R)-3-(Show MoreShow Less
SMILES CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)OShow MoreShow Less

Specifications

Type Streptomycin Sulfate
Loss on Drying 7% max.
Quantity 25 g
Solubility Information Solubility in water: soluble.
Formula Weight 1457.4
Chemical Name or Material Streptomycin sulfate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462621 Certificate TypeCertificate of Analysis Date17/05/2024 Catalog Number455340050,455340250,455341000
Lot NumberA0458703 Certificate TypeCertificate of Analysis Date18/12/2023 Catalog Number455340050,455340250,455341000
Lot NumberA0455487 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number455340050,455340250,455341000
Lot NumberA0452192 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number455340050,455340250,455341000
Lot NumberA0450191 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number455340050,455340250,455341000
compliance-icons compliance-icons
Product Identifier
  • Streptomycin Sulfate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Streptomycin sulfate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Streptomycin sulfate >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.