missing translation for 'onlineSavingsMsg'
Learn More

Streptozotocin, ≥97%, MP Biomedicals™

Product Code. 11437640
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 18883-66-4
Molecular Formula C8H15N3O7
Molecular Weight (g/mol) 265.222
InChI Key ZSJLQEPLLKMAKR-UHEPRFFZSA-N
Synonym STZ, 2-Deoxy-2-(3-methyl-3-nitrosoureido)-D-glucopyranose
PubChem CID 45357367
IUPAC Name 1-methyl-1-nitroso-3-[(2R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]urea
SMILES CN(C(=O)NC1C(C(C(OC1O)CO)O)O)N=O

Specifications

Type Streptozotocin
Physical Form Solid
Assay Percent Range ≥97%
Formula Weight 265.2
Color White
Quantity 1 g
Chemical Name or Material Streptozotocin
compliance-icons
Product Identifier
  • Streptozotocin
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : streptozotocin
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'