missing translation for 'onlineSavingsMsg'
Learn More

Ansell Edmont™ String Knits™ 76-160 Series Natural Cotton Light Weight Gloves

Product Code. 12656971
Click to view available options
Size:
6
7
8
9
10
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Can be worn on their own or as liners. Permits a longer, more comfortable wear with no weak points which might split or fray.

  • Stringknit construction allows good air flow to the hand.
  • Hands don't become hot and sweaty.
  • Ambidextrous: No need to pair, therefore more economical.
  • All styles washable at 40°C.
  • Will not shrink, stiffen or lose their shape.
  • More hygienic for the wearer, better value for the provider.
TRUSTED_SUSTAINABILITY

Specifications

Back Material Polyester/Cotton Blend
Construction Seamless Knit
Cuff Material Polyester/Cotton
Color Natural
Grade 13 gauge
Material Polyester, Cotton
Product Type Light Weight Gloves
Size 7
Style 76-160
Glove Cuff Color Natural
Glove Cuff Style Knitwrist
Lining Polyester/Cotton
Quantity 12 Pairs
Resistant To Mechanical protection
Series 76-160
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'