missing translation for 'onlineSavingsMsg'
Learn More

Ansell Edmont™ String Knits™ 76-160 Series Natural Cotton Light Weight Gloves

Product Code. 12666971
Click to view available options
Size:
6
7
8
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Can be worn on their own or as liners. Permits a longer, more comfortable wear with no weak points which might split or fray.

  • Stringknit construction allows good air flow to the hand.
  • Hands don't become hot and sweaty.
  • Ambidextrous: No need to pair, therefore more economical.
  • All styles washable at 40°C.
  • Will not shrink, stiffen or lose their shape.
  • More hygienic for the wearer, better value for the provider.
TRUSTED_SUSTAINABILITY

Specifications

Back Material Polyester/Cotton Blend
Construction Seamless Knit
Cuff Material Polyester/Cotton
Color Natural
Grade 13 gauge
Material Polyester, Cotton
Product Type Light Weight Gloves
Size 8
Style 76-160
Glove Cuff Color Natural
Glove Cuff Style Knitwrist
Lining Polyester/Cotton
Quantity 12 Pairs
Resistant To Mechanical protection
Series 76-160
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'