missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Stringknits™ 76-200 Series Lightweight Nylon Gloves

Permits longer, more comfortable wear with no weak points which might split or fray. Ansell Stringknits™ 76-200 Series Lightweight Nylon Gloves can be worn on their own or as liners.

£24.50 - £25.48

Specifications

Back Material Nylon
Construction Seamless Knit
Cuff Material Nylon
Color White
Grade 13 gauge
View More Specs

Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
12920135
View Documents
Ansell™
76200070
Small
£24.50
12 pairs
In Stock
to see stock.
Edge
Add to basket
 
12950135
View Documents
Ansell™
76200100
X-Large
£24.50
12 pairs
In Stock
to see stock.
Add to basket
 
12930135
View Documents
Ansell™
76200080
Medium
£25.48
12 pairs
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
 
12940135
View Documents
Ansell™
76200090
Large
£25.48
12 pairs
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
 
Description

Description

  • Good value product protection
  • No seams to rub or irritate
  • Ambidextrous
  • Allows good air flow to the hand, preventing heat and sweat
  • Washable at 40°C without shrinkage or loss of shape
  • Soft, comfortable, flexible and low lint
  • Applications include general handling, delicate manipulation, fine assembly of dry parts
Specifications

Specifications

Nylon
Nylon
13 gauge
Nylon
White
12 Pairs
76-200
Seamless Knit
White
Ambidextrous
76-200
Nylon
Mechanical protection
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell™ Stringknits™ 76-200 Series Lightweight Nylon Gloves

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.