Learn More
Ansell™ String Knits™ 76-202 Series White Nylon Light Weight Gloves
Product Code. 12636981
Click to view available options
Size:
6
7
8
9
10
11
Can be worn on their own or as liners. Permits a longer, more comfortable wear with no weak points which might split or fray.
  • For general handling and delicate manipulation
  • Can also be worn as liners
  • Without seams to rub or irritate
  • Ambidextrous
  • Washable at 40°C
  • Does not shrink, stiffen or lose shape
  • Nylon liner
  • Knitwrist cuff
TRUSTED_SUSTAINABILITY
Specifications
Back Material Nylon
Construction Seamless Knit
Cuff Material Nylon
Product Type Lightweight Gloves
Size 11
Style 76-202
Glove Cuff Color White
Glove Cuff Style Knit
Lining Nylon
Quantity 12 Pairs
Resistant To Mechanical protection
Series 76-202