Learn More
Astell Scientific™ Strip Chart Record
Product Code. 15731449
Click to view available options
Description:
Single Pen
Unit Size:
Each
Product Code. 15731449
Brand: Astell Scientific™ AAR022
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Fisher Scientific - Astell Scientific Strip Chart Record with a single pen for recording chamber temperature only.
  • Microprocessor controlled strip chart recorders
  • Span through the use of accurate stepper motors
  • Incorporates a high definition alphanumeric display, indicating temperature and pressure
Specifications
Description Single Pen