missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Stripping Buffer-2, 4X
£34.80 - £60.10
Specifications
Product Type | Western Blot Stripping Buffer |
---|---|
Form | Liquid |
Concentration | 4X |
Content And Storage | Ambient temperatures |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15404489
|
Thermo Scientific™
J62023.AK |
250 mL |
£34.80
250mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15414489
|
Thermo Scientific™
J62023.AP |
500 mL |
£60.10
500mL |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécification
Western Blot Stripping Buffer | |
Liquid | |
4X | |
Ambient temperatures |