missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Stripping Buffer, 4X
£30.00
Specifications
Product Type | Western Blot Stripping Buffer |
---|---|
Form | Liquid |
Concentration | 4X |
Content And Storage | Ambient temperatures |
Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
15453609
|
Thermo Scientific™
J60925.AK |
250 mL | N/A | ||||||
Descrizione
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifica
Western Blot Stripping Buffer | |
Liquid | |
4X | |
Ambient temperatures |