missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Stripwell™ Microplates
GREENER_CHOICE

Product Code. p-154435 Shop All Corning Products
Click to view available options
Color:
Black
Clear
White
Binding Property:
High Binding
Medium Binding
Total Volume (Metric) Well:
190 μL
360 μL
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 10347891

Brand: Corning™ 3913

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Designed and treated to provide uniform binding, high optical clarity, and low background interference. Corning™ Stripwell™ Microplates provide a superior solution for in vitro diagnostic assays.

Corning™ Stripwell™ microplates may be used in a variety of laboratory assays. The flat bottom strips are designed to easily break apart and are pre-assembled in an “egg-crate” style strip holder that allows each individual well to be positioned back into the microplate once broken.

  • Stripwell microplates have 96-well flat bottom polystyrene format.
  • Low volume and standard Stripwell microplates have well volumes of 190 μL and 360 μL, respectively.
  • 1 x 8 strips are designed to fit only one way into the strip holder, eliminating the chance of misorientation.

Order Info

Bulk packed. Cat. No. 07-200-24 has individual 1 x 8 strips without frame.

Certifications

Cat. Nos. 07-200-31 and -32 have certified surface chemistry.

TRUSTED_SUSTAINABILITY

Specifications

Binding Property Medium Binding
Format Strip Plate
Lid Without Lid
Array 8 x 12
Binding Type Medium Binding
Color White
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 75 to 200 μL
Sterility Non-sterile
Total Volume (Metric) Well 360 μL
Well Shape Round
For Use With (Application) In-vitro diagnostic assays
No. per Case 100/Cs.
No. per Pack 25/Pk.
Quantity 100/Cs.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.