missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STRO-1 Monoclonal Antibody (STRO-1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 398401
304.50 GBP valid until 2025-03-16
Use promo code "24070" to get your promotional price.
This item is not returnable.
View return policy
Description
STRO-1 is a cell surface antigen expressed by stromal elements in human bone marrow. STRO-1 binds to approximately 10% of bone marrow mononuclear cells, greater than 95% of which are nucleated erythroid precursors, but does not react with committed progenitor cells (colony-forming unit granulocyte-macrophage (CFU-GM), erythroid bursts (BFU-E), and mixed colonies (CFU-Mix). From bone marrow cells, the frequency of fibroblast colony forming cells (CFU-F) is enriched approximately 100-fold in the STRO-1-positive/Glycophorin A-negative population than in the STRO-1-positive/Glycophorin A-positive population. When plated under long-term BM culture conditions, STRO-1-positive cells generate adherent cell layers containing multiple stromal cell types, including adipocytes, smooth muscle cells, and fibroblastic elements. A STRO-1-positive enriched subset of marrow cells is capable of differentiating into multiple mesenchymal lineages including hematopoiesis-supportive stromal cells with a vascular smooth muscle-like phenotype, adipocytes, osteoblasts and chondrocytes. STRO-1 is a valuable antibody for the identification, isolation and functional characterization of human bone marrow stromal cell precursors.
Specifications
STRO-1 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.1% sodium azide | |
Mouse | |
100 μL | |
Primary | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Western Blot | |
STRO-1 | |
Unconjugated | |
STRO1 | |
CD34+ human bone marrow cells | |
RUO | |
Human | |
Antibody | |
IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
STRO-1 Monoclonal Antibody (STRO-1), Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction