missing translation for 'onlineSavingsMsg'
Learn More

Strontium for AAS 10g/L HNO3 matrix, Fisher Chemical™

Product Code. 12903375
Click to view available options
Quantity:
100 mL
Packaging:
Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Concentration 10g/L ±0.02g/L
CAS 7440-24-6
Packaging Bottle
Quantity 100 mL
Type Single-element Concentrated Standard Solution
Molecular Formula Sr
UN Number 1760
InChI Key CIOAGBVUUVVLOB-UHFFFAOYSA-N
SMILES [Sr]
IUPAC Name strontium
Molecular Weight (g/mol) 87.62
PubChem CID 5359327
ChEBI CHEBI:33324
Solvent or Matrix 5% HNO3
Physical Form Liquid
Chemical Name or Material Strontium
Show More Show Less
compliance-icons
Product Identifier
  • Single-element Standard Solution for AAS. Strontium (Sr) 10000mg/l in HNO3 5%
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'