missing translation for 'onlineSavingsMsg'
Learn More

Strontium bromide hexahydrate, 95%

Product Code. 11334879
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11334879

Brand: Thermo Scientific Alfa Aesar 072105.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-53-9 | Br2H12O6Sr | 355.518 g/mol

Strontium bromide hexahydrate is used in flares and also used in some pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-53-9
Molecular Formula Br2H12O6Sr
Molecular Weight (g/mol) 355.518
MDL Number MFCD00150361
InChI Key FLMJUJXBFKFYOZ-UHFFFAOYSA-L
Synonym unii-7i32n2ud6w, strontium dibromide hexahydrate, strontium bromide, reagent, strontium 2+ hexahydrate dibromide, strontium 2+ ion hexahydrate dibromide
PubChem CID 165644
IUPAC Name strontium;dibromide;hexahydrate
SMILES O.O.O.O.O.O.[Br-].[Br-].[Sr+2]

Specifications

Physical Form Crystalline
Quantity 500 g
Assay Percent Range 95%
Solubility Information Soluble in water,alcohol
Formula Weight 355.53 (247.44 Anhydrous)
Sensitivity Hygroscopic
Density 2.386
Chemical Name or Material Strontium bromide hexahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.