missing translation for 'onlineSavingsMsg'
Learn More

Strontium bromide hexahydrate, 99%, Thermo Scientific Chemicals

£46.93 - £171.86

Chemical Identifiers

CAS 7789-53-9
Molecular Formula Br2H12O6Sr
Molecular Weight (g/mol) 355.518
MDL Number MFCD00150361
InChI Key FLMJUJXBFKFYOZ-UHFFFAOYSA-L
Synonym unii-7i32n2ud6w, strontium dibromide hexahydrate, strontium bromide, reagent, strontium 2+ hexahydrate dibromide, strontium 2+ ion hexahydrate dibromide
PubChem CID 165644
IUPAC Name strontium;dibromide;hexahydrate
SMILES O.O.O.O.O.O.[Br-].[Br-].[Sr+2]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11495117
View Documents
Thermo Scientific Alfa Aesar
B22553.22
100 g
£46.93
100g
Please to purchase this item. Need a web account? Register with us today!
11405127
View Documents
Thermo Scientific Alfa Aesar
B22553.36
500 g
£171.86
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Strontium bromide hexahydrate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Strontium bromide hexahydrate is used as pharmaceutical intermediate.

Solubility
Soluble in water (2850 g/L at 20°C).

Notes
m.p. 88° (rapidly heated) , 180° -6H{2}O
Specifications

Chemical Identifiers

7789-53-9
355.518
FLMJUJXBFKFYOZ-UHFFFAOYSA-L
165644
O.O.O.O.O.O.[Br-].[Br-].[Sr+2]
Br2H12O6Sr
MFCD00150361
unii-7i32n2ud6w, strontium dibromide hexahydrate, strontium bromide, reagent, strontium 2+ hexahydrate dibromide, strontium 2+ ion hexahydrate dibromide
strontium;dibromide;hexahydrate

Specifications

7789-53-9
SrBr2·6H2O
14,8837
Soluble in water (2850g/L at 20°C).
O.O.O.O.O.O.[Br-].[Br-].[Sr+2]
355.518
355.53 (247.44 Anhydrous)
Odorless
2.386
Br2H12O6Sr
MFCD00150361
unii-7i32n2ud6w, strontium dibromide hexahydrate, strontium bromide, reagent, strontium 2+ hexahydrate dibromide, strontium 2+ ion hexahydrate dibromide
FLMJUJXBFKFYOZ-UHFFFAOYSA-L
strontium;dibromide;hexahydrate
165644
99%
Hygroscopic
Strontium bromide hexahydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 233-969-5

missing translation for 'rtecsNumber' : WK8225000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only