missing translation for 'onlineSavingsMsg'
Learn More
Strontium bromide, ultra dry, 99.995% (metals basis), Thermo Scientific Chemicals Product Code.: 11368567

Strontium bromide, ultra dry, 99.995% (metals basis), Thermo Scientific Chemicals

Product Code. 11368567
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11368567

Brand: Thermo Scientific Alfa Aesar 035717.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used as sedatives in medicine and analytical reagents, for facile synthesis of poly(phenylcarbyne), which is a precursor to diamond-like carbon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10476-81-0
Molecular Formula Br2Sr
Molecular Weight (g/mol) 247.428
MDL Number MFCD00049548
InChI Key YJPVTCSBVRMESK-UHFFFAOYSA-L
Synonym acmc-20ajmt, strontium bromide srbr2, strontium 2+ ion dibromide
PubChem CID 5360351
IUPAC Name strontium;dibromide
SMILES [Br-].[Br-].[Sr+2]

Specifications

Melting Point 643°C
Color White
Physical Form Powder
Quantity 5 g
Linear Formula SrBr2
Merck Index 14,8837
Solubility Information Soluble in water,carbinol,ethanol,amyl alcohol. Insoluble in ether.
Formula Weight 247.44
Percent Purity 99.995%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 4.216 g/mL
Assay (metals basis)
Chemical Name or Material Strontium bromide, Ultra dry
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT18I046 Certificate TypeCertificate of Analysis Date08/08/2022 Catalog Number035717.03,035717.06
Lot NumberT18I045 Certificate TypeCertificate of Analysis Date06/08/2022 Catalog Number035717.03,035717.06
Lot NumberR09F020 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number035717.03,035717.06
Lot NumberW16F045 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number035717.03,035717.06
Lot NumberQ10E038 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number035717.03,035717.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Strontium bromide, ultra dry, 99.995% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.