missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strontium carbonate, 97.5%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
5 kg
Description
As a catalyst, in radiation-resistant glass, in ceramic ferrites, and in pyrotechnicsUsed as a colorant in fireworks. It is also used in ceramics industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 1100°C (decomposition) |
Physical Form | Powder |
Quantity | 250 g |
Assay Percent Range | 97.5% |
Solubility Information | Practically insoluble in water. Soluble in acids and carbonated waterSoluble in ammonium chloride. Slightly soluble in ammonia and water. |
Formula Weight | 147.63 |
Density | 3.7 |
Chemical Name or Material | Strontium carbonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'