missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strontium Carbonate, Extra Pure, SLR, Fisher Chemical™
£57.30 - £163.00
Chemical Identifiers
CAS | 1633-05-2 |
---|---|
Molecular Formula | CO3Sr |
Molecular Weight (g/mol) | 147.63 |
MDL Number | MFCD00011250 |
InChI Key | LEDMRZGFZIAGGB-UHFFFAOYSA-L |
Synonym | strontium carbonate, strontianite, carbonic acid, strontium salt 1:1, unii-41ypu4mmca, strontium carbonate srco3, ccris 3203, carbonic acid strontium salt 1:1, 41ypu4mmca, strontium, reference standard solution, strontiumcarbonateShow More |
PubChem CID | 15407 |
IUPAC Name | strontium;carbonate |
SMILES | [Sr++].[O-]C([O-])=O |
Description
Chemical Identifiers
1633-05-2 | |
147.63 | |
LEDMRZGFZIAGGB-UHFFFAOYSA-L | |
15407 | |
[Sr++].[O-]C([O-])=O |
CO3Sr | |
MFCD00011250 | |
strontium carbonate, strontianite, carbonic acid, strontium salt 1:1, unii-41ypu4mmca, strontium carbonate srco3, ccris 3203, carbonic acid strontium salt 1:1, 41ypu4mmca, strontium, reference standard solution, strontiumcarbonateShow More | |
strontium;carbonate |
Specifications
1633-05-2 | |
Solid | |
MFCD00011250 | |
LEDMRZGFZIAGGB-UHFFFAOYSA-L | |
strontium;carbonate | |
15407 | |
Plastic powder jar |
White | |
CO3Sr | |
strontium carbonate, strontianite, carbonic acid, strontium salt 1:1, unii-41ypu4mmca, strontium carbonate srco3, ccris 3203, carbonic acid strontium salt 1:1, 41ypu4mmca, strontium, reference standard solution, strontiumcarbonate | |
[Sr++].[O-]C([O-])=O | |
147.63 | |
147.6182g/mol |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Strontium Carbonate, Extra Pure, SLR, Fisher Chemical™