missing translation for 'onlineSavingsMsg'
Learn More
Strontium carbonate, Puratronic™, 99.994% (metals basis), Thermo Scientific Chemicals Product Code.: 11388058

Strontium carbonate, Puratronic™, 99.994% (metals basis), Thermo Scientific Chemicals

Product Code. 11388058
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11388058

Brand: Thermo Scientific Alfa Aesar 036618.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used as a colorant in fireworks. It is also used in ceramics industry.

Used as a colorant in fireworks. It is also used in ceramics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1633-05-2
Molecular Formula CO3Sr
Molecular Weight (g/mol) 147.63
MDL Number MFCD00011250
InChI Key LEDMRZGFZIAGGB-UHFFFAOYSA-L
Synonym strontium carbonate, strontianite, carbonic acid, strontium salt 1:1, unii-41ypu4mmca, strontium carbonate srco3, ccris 3203, carbonic acid strontium salt 1:1, 41ypu4mmca, strontium, reference standard solution, strontiumcarbonateShow MoreShow Less
PubChem CID 15407
IUPAC Name strontium;carbonate
SMILES [Sr++].[O-]C([O-])=O

Specifications

Melting Point 1100°C (decomposition)
Physical Form Powder
Quantity 25 g
Solubility Information Soluble in ammonium chloride. Slightly soluble in ammonia and water.
Formula Weight 147.63
Percent Purity 99.994%
Packaging Plastic bottle
Specific Gravity 3.7
Density 3.7
Chemical Name or Material Strontium carbonate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN15J007 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number036618.14
Lot NumberQ25J018 Certificate TypeCertificate of Analysis Date01/05/2023 Catalog Number036618.14
Lot NumberR03I062 Certificate TypeCertificate of Analysis Date24/05/2022 Catalog Number036618.14,036618.36,036618.22
Lot NumberP23H012 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number036618.14,036618.36,036618.22
Lot Number24040 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number036618.14,036618.36,036618.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Strontium carbonate, Puratronic™, 99.994% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.