missing translation for 'onlineSavingsMsg'
Learn More
Strontium chloride hexahydrate, 99+%, ACS reagent, Thermo Scientific Chemicals Product Code.: 10349220

Strontium chloride hexahydrate, 99+%, ACS reagent, Thermo Scientific Chemicals

Product Code. 10349220
100 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10349220

Brand: Thermo Scientific Acros 315081000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-70-4
Molecular Formula Cl2H12O6Sr
Molecular Weight (g/mol) 266.61
MDL Number MFCD00149865
InChI Key AMGRXJSJSONEEG-UHFFFAOYSA-L
IUPAC Name strontium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Sr++]

Specifications

Melting Point 115.0°C
Color White
pH 5.0 to 7.0 (5% soln. at 25°C)
Physical Form Crystals
Quantity 100 g
Assay Percent Range 99.0 to 103% (Complexometry)
Linear Formula SrCl2·6H2O
Merck Index 15, 8970
Solubility Information Solubility in water: 106.2 g/100mL (0°C). Other solubilities: soluble in alcohol
Formula Weight 266.62
Percent Purity ≥99%
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Strontium chloride hexahydrate, For analysis ACS
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467524 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number315081000,315085000
Lot NumberA0464137 Certificate TypeCertificate of Analysis Date10/07/2024 Catalog Number315081000,315085000
Lot NumberA0460307 Certificate TypeCertificate of Analysis Date16/02/2024 Catalog Number315081000,315085000
Lot NumberA0457684 Certificate TypeCertificate of Analysis Date10/11/2023 Catalog Number315081000,315085000
Lot NumberA0449950 Certificate TypeCertificate of Analysis Date01/02/2023 Catalog Number
compliance-icons compliance-icons
Product Identifier
  • Strontium chloride hexahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Strontium chloride hexahydrate, 99+%, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.