Learn More
Strontium chloride hexahydrate, 98%, Thermo Scientific Chemicals
£31.36 - £98.99
Chemical Identifiers
CAS | 10025-70-4 |
---|---|
Molecular Formula | Cl2H12O6Sr |
Molecular Weight (g/mol) | 266.61 |
MDL Number | MFCD00149865 |
InChI Key | AMGRXJSJSONEEG-UHFFFAOYSA-L |
IUPAC Name | strontium(2+) hexahydrate dichloride |
SMILES | O.O.O.O.O.O.[Cl-].[Cl-].[Sr++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11472778
|
Thermo Scientific Alfa Aesar
A16764.30 |
250 g |
£31.36
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11462778
|
Thermo Scientific Alfa Aesar
A16764.0B |
1000 g |
£98.99
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Strontium chloride hexahydrate is a precursor to prepare strontium chromate which is used as a corrosion inhibitor for aluminum. It is involved in the synthesis of cadmium sulfide core photocatalytic nanoparticles. It is used as a red coloring agent in pyrotechnics. Furthermore, it is utilized in glass-making and metallurgy. In the medical field, its radioactive isotope strontium-89 is used for the treatment of bone cancer. It acts as a catalyst in the conversion of ketones and aldehydes to the corresponding gem-dihydroperoxides using aqueous hydrogen peroxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsStrontium chloride hexahydrate is a precursor to prepare strontium chromate which is used as a corrosion inhibitor for aluminum. It is involved in the synthesis of cadmium sulfide core photocatalytic nanoparticles. It is used as a red coloring agent in pyrotechnics. Furthermore, it is utilized in glass-making and metallurgy. In the medical field, its radioactive isotope strontium-89 is used for the treatment of bone cancer. It acts as a catalyst in the conversion of ketones and aldehydes to the corresponding gem-dihydroperoxides using aqueous hydrogen peroxide.
Solubility
Soluble in water. Slightly soluble in ethanol and acetone. Insoluble in ammonia.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
10025-70-4 | |
266.61 | |
AMGRXJSJSONEEG-UHFFFAOYSA-L | |
O.O.O.O.O.O.[Cl-].[Cl-].[Sr++] |
Cl2H12O6Sr | |
MFCD00149865 | |
strontium(2+) hexahydrate dichloride |
Specifications
10025-70-4 | |
MFCD00149865 | |
Soluble in water. Slightly soluble in ethanol and acetone. Insoluble in ammonia. | |
O.O.O.O.O.O.[Cl-].[Cl-].[Sr++] | |
266.61 | |
≥99% | |
Hygroscopic | |
Strontium chloride hexahydrate |
Cl2H12O6Sr | |
14,8840 | |
AMGRXJSJSONEEG-UHFFFAOYSA-L | |
strontium(2+) hexahydrate dichloride | |
266.62 (158.53 Anhydrous) | |
1.65 | |
1.93 |
Safety and Handling
GHS H Statement
H318
Causes serious eye damage.
P261-P271-P280-P304+P340-P305+P351+P338-P310-P312-P501c
H318-H335
missing translation for 'einecsNumber' : 233-971-6
missing translation for 'rtecsNumber' : WK8575000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Strontium chloride hexahydrate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Strontium chloride (SrCl2), hexahydrate
RUO – Research Use Only