missing translation for 'onlineSavingsMsg'
Learn More

Strontium chloride hexahydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals

£26.73 - £66.23

Chemical Identifiers

CAS 10025-70-4
Molecular Formula Cl2H12O6Sr
Molecular Weight (g/mol) 266.61
MDL Number MFCD00149865
InChI Key AMGRXJSJSONEEG-UHFFFAOYSA-L
IUPAC Name strontium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Sr++]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11327728
View Documents Promotion Details
Thermo Scientific Alfa Aesar
033393.22
100 g
£26.73
100g
Please to purchase this item. Need a web account? Register with us today!
11337728
View Documents Promotion Details
Thermo Scientific Alfa Aesar
033393.36
500 g
£66.23
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Strontium chloride hexahydrate is a precursor to prepare strontium chromate which is used as a corrosion inhibitor for aluminum. It is involved in the synthesis of cadmium sulfide core photocatalytic nanoparticles. It is used as a red coloring agent in pyrotechnics. Furthermore, it is utilized in glass-making and metallurgy. In the medical field, its radioactive isotope strontium-89 is used for the treatment of bone cancer. It acts as a catalyst in the conversion of ketones and aldehydes to the corresponding gem-dihydroperoxides using aqueous hydrogen peroxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10025-70-4
266.61
AMGRXJSJSONEEG-UHFFFAOYSA-L
O.O.O.O.O.O.[Cl-].[Cl-].[Sr++]
Cl2H12O6Sr
MFCD00149865
strontium(2+) hexahydrate dichloride

Specifications

10025-70-4
99.0 to 103.0%
MFCD00149865
Soluble in water. Slightly soluble in ethanol and acetone. Insoluble in ammonia.
O.O.O.O.O.O.[Cl-].[Cl-].[Sr++]
266.61
99.0 to 103.0%
1.65
1.93 g/mL
Granules
Cl2H12O6Sr
14,8840
AMGRXJSJSONEEG-UHFFFAOYSA-L
strontium(2+) hexahydrate dichloride
266.62 (158.53 Anhydrous)
ACS
Hygroscopic
Strontium chloride hexahydrate, For analysis ACS
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318

P261-P271-P280-P304+P340-P305+P351+P338-P310-P312-P501c

H318-H335

missing translation for 'einecsNumber' : 233-971-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Strontium chloride hexahydrate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Strontium chloride hexahydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only