missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strontium Chloride Hexahydrate, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 g
Specifications
Specifications
Melting Point | 115°C |
Color | White |
pH | 5.0 to 7.0 |
Physical Form | Solid |
Quantity | 500 g |
Formula Weight | 266.62g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Strontium chloride hexahydrate
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'