missing translation for 'onlineSavingsMsg'
Learn More

Strontium chloride hexahydrate, Puratronic™, 99.9965% (metals basis), Thermo Scientific Chemicals

Catalog Number 11397037
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
10025-70-4
Cl2H12O6Sr
266.61
MFCD00149865
AMGRXJSJSONEEG-UHFFFAOYSA-L
strontium(2+) hexahydrate dichloride
O.O.O.O.O.O.[Cl-].[Cl-].[Sr++]
This item is not returnable. View return policy
10025-70-4
Cl2H12O6Sr
266.61
MFCD00149865
AMGRXJSJSONEEG-UHFFFAOYSA-L
strontium(2+) hexahydrate dichloride
O.O.O.O.O.O.[Cl-].[Cl-].[Sr++]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Powder
Quantity 25 g
Merck Index 14,8840
Formula Weight 266.62 (158.53 Anhydrous)
Percent Purity 99.9965%
Grade Puratronic™
Refractive Index 1.65
Sensitivity Hygroscopic
Density 1.93 g/mL
Assay (metals basis)
Chemical Name or Material Strontium chloride hexahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'