missing translation for 'onlineSavingsMsg'
Learn More
Strontium, distilled dendritic pieces, 99.95% (metals basis), Thermo Scientific Chemicals Product Code.: 11338947

Strontium, distilled dendritic pieces, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11338947
5 g, Ampule
Click to view available options
Quantity:
5 g
Packaging:
Ampule
This item is not returnable. View return policy

Product Code. 11338947

Brand: Thermo Scientific Alfa Aesar 038624.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-24-6
Molecular Formula Sr
MDL Number MFCD00134060
UN Number UN3208
Quantity 5 g
Synonym strontium, elemental, unii-yzs2rpe8le, yzs2rpe8le, estroncio, granules, 19mm 0.76in & down metals basis, strontium, distilled dendritic pieces, atom, pieces, strontium, chunks, pieces, dendritic
InChI Key CIOAGBVUUVVLOB-UHFFFAOYSA-N
SMILES [Sr]
IUPAC Name strontium
Molecular Weight (g/mol) 87.62
PubChem CID 5359327
ChEBI CHEBI:33324
Percent Purity 99.95%
Packaging Ampule
Sensitivity Air and moisture sensitive
Assay (metals basis)
Chemical Name or Material Strontium, Distilled dendritic pieces
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ23H017 Certificate TypeCertificate of Analysis Date24/02/2025 Catalog Number038624.06
Lot NumberY15K041 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog Number038624.06
Lot NumberY15K042 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog Number038624.06
Lot NumberY15K040 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog Number038624.06
Lot NumberX21G040 Certificate TypeCertificate of Analysis Date09/11/2023 Catalog Number038624.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Strontium, distilled dendritic pieces, 99.95% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.