missing translation for 'onlineSavingsMsg'
Learn More

Strontium dodecairon nonadecaoxide, 99.0% (metals basis)

Product Code. p-7049774
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11348067

Brand: Thermo Scientific Alfa Aesar 022569.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12023-91-5 | Fe2O5Sr2 | 366.93 g/mol

Strontium dodecairon nonadecaoxide is used in manufacture of mixtures, granulates for plastic articles, production of magnet articles by compacting, sintering or other processes. And it is also used as an additive to flares, pyrotechnics, provides high refractive index to glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12023-91-5
Molecular Formula Fe2O5Sr2
Molecular Weight (g/mol) 366.93
MDL Number MFCD00075632
InChI Key GKHGYOYJUWWDJG-UHFFFAOYSA-N
Synonym strontium ferrite, strontiumferrit, strontium dodecairon nonadecaoxide, oxo oxoferriooxy iron;oxostrontium, strontium ferrite, powder, hexakis ferric oxide ; strontium oxide, strontium ferrite, nanopowder, <100 nm particle size bet trace metals basis
PubChem CID 16213227
IUPAC Name oxo(oxoferriooxy)iron;oxostrontium
SMILES [O--].[O--].[O--].[O--].[O--].[Fe+3].[Fe+3].[Sr++].[Sr++]

Specifications

Melting Point >450°C
Physical Form Powder
Quantity 5 g
Solubility Information Soluble in organic solvents.
Formula Weight 1061.77
Percent Purity 99.5%
Assay (metals basis)
Chemical Name or Material Strontium dodecairon nonadecaoxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.