missing translation for 'onlineSavingsMsg'
Learn More
Strontium Fluoride, 99.99% (Metal Basis), Thermo Scientific Chemicals, Puratronic™ Product Code.: 11327047

Strontium Fluoride, 99.99% (Metal Basis), Thermo Scientific Chemicals, Puratronic™

Product Code. 11327047
5 g
Click to view available options
Quantity:
0.5 g
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11327047

Brand: Thermo Scientific Alfa Aesar 010878.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Strontium fluoride is used as an optical coating on lenses, and as a thermoluminescent dosimeter crystal. It also finds use as a water insoluble strontium source for use in oxygen-sensitive applications such as metal production. It serves both as a source of strontium and fluoride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-48-4
Molecular Formula F2Sr
Molecular Weight (g/mol) 125.62
MDL Number MFCD00011251
InChI Key FVRNDBHWWSPNOM-UHFFFAOYSA-L
Synonym strontium difluoride, unii-efy8gjs81z, strontium fluoride srf2, efy8gjs81z, strontium 2+ ion difluoride, fluoride, sr, fluoride, strontium 2+ difluoride, beta-bromo isovaleric acid, ksc379a4r, strontium fluoride 250gShow MoreShow Less
PubChem CID 82210
SMILES [F-].[F-].[Sr++]

Specifications

Melting Point 1473°C
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.99%, metal basis
Solubility Information Insoluble in water.
IUPAC Name strontium(2+) difluoride
Formula Weight 125.62
Refractive Index 1.442
Density 4.24
Chemical Name or Material Strontium Fluoride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN15J021 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number010878.06,010878.14,010878.22
Lot NumberP30I038 Certificate TypeCertificate of Analysis Date11/08/2022 Catalog Number010878.06,010878.14,010878.22
Lot NumberQ21I013 Certificate TypeCertificate of Analysis Date11/08/2022 Catalog Number010878.06,010878.14,010878.22
Lot Number23016 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010878.06,010878.14,010878.22
Lot Number23568 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010878.06,010878.14,010878.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Strontium Fluoride, 99.99% (Metal Basis), Thermo Scientific Chemicals, Puratronic™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.