missing translation for 'onlineSavingsMsg'
Learn More
Strontium fluoride, 99% min, Thermo Scientific Chemicals Product Code.: 11336608

Strontium fluoride, 99% min, Thermo Scientific Chemicals

Product Code. 11336608
1 kg
Click to view available options
Quantity:
250 g
1 kg
5 kg
This item is not returnable. View return policy

Product Code. 11336608

Brand: Thermo Scientific Alfa Aesar 012201.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Strontium fluoride is used as an optical material for a small range of special applications. It also used as use is as a carrier of strontium-90 radioisotope in radioisotope thermoelectric generators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-48-4
Molecular Formula F2Sr
Molecular Weight (g/mol) 125.62
MDL Number MFCD00011251
InChI Key FVRNDBHWWSPNOM-UHFFFAOYSA-L
Synonym strontium difluoride, unii-efy8gjs81z, strontium fluoride srf2, efy8gjs81z, strontium 2+ ion difluoride, fluoride, sr, fluoride, strontium 2+ difluoride, beta-bromo isovaleric acid, ksc379a4r, strontium fluoride 250gShow MoreShow Less
PubChem CID 82210
IUPAC Name strontium;difluoride
SMILES [F-].[F-].[Sr++]

Specifications

Melting Point 1473°C
Color White
Physical Form Powder
Quantity 1 kg
Merck Index 14,8842
Solubility Information Slightly soluble in water (11;7mg at 18°C/100mL), dilute acids; Decomposes in strong acids
Formula Weight 125.62
Percent Purity ≤99%
Odor Odorless
Refractive Index 1.442
Density 4.240 g/mL
Chemical Name or Material Strontium fluoride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY01K010 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog Number012201.A7,012201.30,012201.A1
Lot NumberW25K009 Certificate TypeCertificate of Analysis Date03/10/2024 Catalog Number012201.A7,012201.30,012201.A1
Lot NumberP14K023 Certificate TypeCertificate of Analysis Date20/03/2024 Catalog Number012201.A7,012201.30,012201.A1
Lot NumberJ10N41 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012201.A7,012201.30,012201.A1
Lot NumberD19X001 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012201.A7,012201.30,012201.A1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Strontium fluoride, 99% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.