missing translation for 'onlineSavingsMsg'
Learn More

Strontium hexafluoro-2,4-pentanedionate, Thermo Scientific™

Product Code. 11307018
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121012-89-3
Molecular Formula C10H2F12O4Sr
Molecular Weight (g/mol) 501.72
MDL Number MFCD00064760
InChI Key KEVXBFHXVKDJRZ-PAMPIZDHSA-L
Synonym strontium hexafluoroacetylacetonate, strontium hexafluoro-2,4-pentanedionate, strontium hexafluoropentanedionate hydrate, 2,4-pentanedione,1,1,1,5,5,5-hexafluoro-,ion 1-,strontium 2:1, 3z-1,1,1,5,5,5-hexafluoro-4-2z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-yl oxy strontio oxy pent-3-en-2-one
PubChem CID 91873267
SMILES [Sr++].[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F.[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F

Specifications

Boiling Point 220°C (0.02mmHg)(sublimation)
Melting Point ∼260°C (decomposition)
Color White
Physical Form Crystalline
Quantity 5 g
IUPAC Name strontium;(Z)-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate
Formula Weight 501.75
Chemical Name or Material Strontium hexafluoro-2,4-pentanedionate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'