missing translation for 'onlineSavingsMsg'
Learn More
Strontium hydroxide octahydrate, 99% (metals basis), Thermo Scientific Chemicals Product Code.: 11354058

Strontium hydroxide octahydrate, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 11354058
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11354058

Brand: Thermo Scientific Alfa Aesar 041351.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Strontium hydroxide octahydrate is used in the sugar industry for its purification, oil and paint industry to improve its drying properties and as an active ingredient in depilatory hair removal products. It is also used as a raw material for manufacturing of other products of strontium salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1311-10-0
Molecular Formula H18O10Sr
Molecular Weight (g/mol) 265.75
MDL Number MFCD00150395
InChI Key UJPWWRPNIRRCPJ-UHFFFAOYSA-L
IUPAC Name strontium(2+) octahydrate dihydroxide
SMILES O.O.O.O.O.O.O.O.[OH-].[OH-].[Sr++]

Specifications

Melting Point 100°C-8H2O
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 99% (metals basis)
Solubility Information Soluble in hot water and acid. Insoluble in acetone.
Formula Weight 265.76 (121.60 Anhydrous)
Sensitivity Air sensitive; Hygroscopic
Density 1.900 g/mL
Chemical Name or Material Strontium hydroxide octahydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN16I054 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number041351.22,041351.36
Lot NumberX30E044 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number041351.22,041351.36
Lot NumberR15D033 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number041351.22,041351.36
Lot NumberG15P38 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number041351.22,041351.36
Lot NumberH16R032 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number041351.22,041351.36
compliance-icons
Product Identifier
  • Strontium hydroxide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Strontium hydroxide,octahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Strontium hydroxide octahydrate, 99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.