missing translation for 'onlineSavingsMsg'
Learn More

Strontium hydroxide octahydrate, Technical grade, Thermo Scientific Chemicals

Catalog Number 11306608
Click to view available options
Quantity:
50 g
500 g
2 kg
10 kg
This item is not returnable. View return policy
1311-10-0
H18O10Sr
265.75
MFCD00150395
UJPWWRPNIRRCPJ-UHFFFAOYSA-L
strontium(2+) octahydrate dihydroxide
O.O.O.O.O.O.O.O.[OH-].[OH-].[Sr++]
This item is not returnable. View return policy
1311-10-0
H18O10Sr
265.75
MFCD00150395
UJPWWRPNIRRCPJ-UHFFFAOYSA-L
strontium(2+) octahydrate dihydroxide
O.O.O.O.O.O.O.O.[OH-].[OH-].[Sr++]

Strontium hydroxide octahydrate is used in the sugar industry for its purification, oil and paint industry to improve its drying properties and as an active ingredient in depilatory hair removal products. It is also used as a raw material for manufacturing of other products of strontium salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 100°C-8H2O
Physical Form Crystalline
Quantity 50 g
Solubility Information Soluble in hot water and acid. Insoluble in acetone.
Formula Weight 265.76 (121.60 Anhydrous)
Grade Technical
Sensitivity Air sensitive; Hygroscopic
Density 1.9
Chemical Name or Material Strontium hydroxide octahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'