missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strontium iodide, anhydrous, 99% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 515°C |
Color | Brown to Yellow |
Physical Form | Powder |
Quantity | 5 g |
Merck Index | 14,8844 |
Formula Weight | 341.43 |
Percent Purity | 97% |
Odor | Odorless |
Sensitivity | Light Sensitive & Hygroscopic |
Density | 4.550g/mL |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Z11J069 | Certificate of Analysis | 08/01/2024 | 047268.22,047268.06,047268.14 |
S20J054 | Certificate of Analysis | 28/06/2023 | 047268.22,047268.06,047268,047268.14 |
S20J066 | Certificate of Analysis | 28/06/2023 | 047268.22,047268.06,047268,047268.14 |
S21J044 | Certificate of Analysis | 27/06/2023 | 047268.22,047268.06,047268,047268.14 |
X28H011 | Certificate of Analysis | 12/04/2022 | 047268.22,047268.06,047268.14 |
Safety and Handling


Product Identifier
- Strontium iodide
- Warning
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P390-Absorb spillage to prevent material damage.
- MIXTURE LIST-Contains : Strontium iodide
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Strontium iodide, anhydrous, 99% (metals basis), Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction