missing translation for 'onlineSavingsMsg'
Learn More
Strontium iodide, anhydrous, 99% (metals basis), Thermo Scientific Chemicals Product Code.: 15482937

Strontium iodide, anhydrous, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 15482937
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 15482937

Brand: Thermo Scientific Alfa Aesar 047268.22

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10476-86-5
MDL Number MFCD00049557

Specifications

Melting Point 515°C
Color Brown to Yellow
Physical Form Powder
Quantity 100 g
Merck Index 14,8844
Formula Weight 341.43
Percent Purity 97%
Odor Odorless
Sensitivity Light Sensitive & Hygroscopic
Density 4.550g/mL
Assay (metals basis)
Chemical Name or Material Strontium iodide, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Strontium iodide, anhydrous, 99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.