missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strontium molybdenum oxide, 99%
Click to view available options
Quantity:
10 g
50 g
250 g
1 kg
Unit Size:
10g
1kg
250g
50g
Description
It is used as an optical material for a small range of special applications, for example, as an optical coating on lenses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | (decomposition) |
| Color | Colorless |
| Physical Form | Powder |
| Quantity | 10 g |
| Assay Percent Range | 99.9% (excluding Ba) |
| Solubility Information | Insoluble in water. Soluble in acid |
| Formula Weight | 247.56 |
| Density | 4.54 |
| Chemical Name or Material | Strontium molybdenum oxide, Ba < 0.3% |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction