Learn More
Strontium nitrate, 99+%, for analysis, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 570.0°C |
Color | White |
pH | 5 to 7 (5% soln. at 20°C) |
Physical Form | Crystalline Powder and/or Crystals |
Quantity | 1 kg |
Assay Percent Range | 99% min. (Complexometry) |
Linear Formula | Sr(NO3)2 |
Merck Index | 15, 8975 |
Solubility Information | Solubility in water: 660g/L (20°C). Other solubilities: slightly soluble in alcohol and acetone |
IUPAC Name | strontium(2+) dinitrate |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0461101 | Certificate of Analysis | 19/03/2024 | 222560010,222562500 |
A0460764 | Certificate of Analysis | 08/03/2024 | 222560010,222562500 |
A0455385 | Certificate of Analysis | 25/08/2023 | 222560010,222562500 |
A0448508 | Certificate of Analysis | 17/08/2023 | 222560010,222562500 |
A0394344 | Certificate of Analysis | 12/12/2022 | 222560010,222562500 |
Safety and Handling


- Strontium nitrate
- Danger
- Serious eye damage/eye irritation Category 1
- Oxidising solid Category 3
- H318-Causes serious eye damage.
- H272-May intensify fire; oxidiser.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.