missing translation for 'onlineSavingsMsg'
Learn More

Strontium nitrate, 99+%, for analysis

Product Code. p-41297 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 10155040

Brand: Thermo Scientific Acros 222562500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Strontium nitrate, 99+%, N2O6Sr, CAS Number-10042-76-9, bdg873aqzl, strontium dinitrate, nitric acid, strontium salt 2:1, hsdb 787, nitric acid, strontium salt, nitrate de strontium french, strontium nitrate sr no3 2, strontium nitrate, strontium ii nitrate 1:2, unii-bdg873aqzl, 1kg | CAS: 10042-76-9 | N2O6Sr | 211.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10042-76-9
Molecular Formula N2O6Sr
Molecular Weight (g/mol) 211.63
MDL Number MFCD00011248
InChI Key DHEQXMRUPNDRPG-UHFFFAOYSA-N
Synonym strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1
PubChem CID 24848
SMILES [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 570.0°C
Color White
pH 5 to 7 (5% soln. at 20°C)
Physical Form Crystalline Powder and/or Crystals
Quantity 250 g
Assay Percent Range 99% min. (Complexometry)
Linear Formula Sr(NO3)2
Merck Index 15, 8975
Solubility Information Solubility in water: 660g/L (20°C). Other solubilities: slightly soluble in alcohol and acetone
IUPAC Name strontium;dinitrate
Formula Weight 211.63
Percent Purity ≥99%
Grade Analytical
Loss on Drying 0.1% max. (105°C)
Packaging Plastic bottle
Density 2.99
Chemical Name or Material Strontium nitrate, p.a.
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Strontium nitrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.